Potassium sulphide, anhydrous ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Dipotassium sulphide
19-3900-5G
19-3900-1G
101442-456EA
369.42
USD
101442-456
101442-454
Potassium sulphide, anhydrous ≥95%
Potassium sulphide
Formula:
K₂S MW: 110.26 Melting Pt: 840 Density: 1.74 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00043083 CAS Number: 1312-73-8 EINECS: 215-197-0 UN: 1382 ADR: 4.2 II Merck Index: 12,07846 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...