1,5-Bis(diphenylphosphino)pentane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

15-0430-5G 15-0430-1G
101418-568EA 225.05 USD
101418-568 101418-566
1,5-Bis(diphenylphosphino)pentane ≥98%
1,5-Bis(diphenylphosphino)pentane

Phosphine, Pincer Ligands and Complexes

Formula: C₂₉H₃₀P₂
MW: 440.5 g/mol
Boiling Pt: 230 °C (0.1 -rr)
Melting Pt: 43 - 46 °C
MDL Number: MFCD00003052
CAS Number: 27721-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR