2-(Diphenylphosphino)benzoic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0133-5G 15-0133-1G
101418-528EA 271.75 USD
101418-528 101418-526
2-(Diphenylphosphino)benzoic acid ≥97%
2-(Diphenylphosphino)benzoic acid

Phosphine

Formula: C₁₉H₁₅O₂P
MW: 306.3 g/mol
Melting Pt: 174…181 °C
MDL Number: MFCD00674024
CAS Number: 17261-28-8
EINECS: 241-293-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR