Sodium triacetoxyborohydride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

11-0595-100G 11-0595-25G
101433-072EA 208.06 USD
101433-072 101433-074
Sodium triacetoxyborohydride ≥95%
Sodium triacetoxyborohydride
Formula: C₆H₁₀BNaO₆
Melting Pt: 106 °C
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 1409
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR