(S,S)-(+)-3,6-Octanediol ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: (3S,6S)-Octane-3,6-diol, (S,S)-(+)-3,6-Dihydroxy octane, (3S,6S)-(+)-3,6-Octanediol

08-2038-250MG 08-2038-1G
101324-522EA 196.74 USD
101324-522 101324-524
(S,S)-(+)-3,6-Octanediol ≥99%
(S,S)-(+)-3,6-Octanediol
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Melting Pt: 49…51 °C
MDL Number: MFCD01631088
CAS Number: 136705-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR