(R,R)-(-)-3,6-Octanediol ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
(3R,6R)-Octane-3,6-diol, (R,R)-(-)-3,6-Dihydroxy octane, (3R,6R)-(-)-3,6-Octanediol
08-2037-1G
08-2037-250MG
101324-520EA
576.06
USD
101324-520
101324-518
(R,R)-(-)-3,6-Octanediol ≥99%
(R,R)-(-)-3,6-Octanediol
Formula:
C₈H₁₈O₂ MW: 146.23 g/mol Melting Pt: 49…51 °C |
MDL Number:
MFCD01631087 CAS Number: 129619-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...