(S,S)-(+)-2,4-Pentanediol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2S,4S)-2,4-Pentanediol, (S,S)-(+)-2,4-Dihydroxy pentane, (2S,4S)-(+)-Pentanediol, (2S,4S)-(+)-2,4-Pentanediol

08-2031-250MG 08-2031-1G
101418-452EA 127.38 USD
101418-452 101418-454
(S,S)-(+)-2,4-Pentanediol ≥99%
(S,S)-(+)-2,4-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 111…113 °C (19 mmHg)
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00064276
CAS Number: 72345-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR