meso-Tetraphenylporphine chlorin-free

Supplier: STREM CHEMICALS INC MS

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

07-2170-50MG 07-2170-250MG
100201-356EA 38.22 USD
100201-356 100206-246
meso-Tetraphenylporphine chlorin-free
meso-Tetraphenylporphine

Porphine

Formula: C₄₄H₃₀N₄
Density: 1.276 g/cm³ (20 °C)
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR