4,4-Dimethyl-2-pentyne ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
t-Butylmethylacetylene, tert-Butyl methyl acetylene, 4,4-Dimethylpent-2-yne
06-0150-5G
06-0150-1G
101324-496EA
281.66
USD
101324-496
101324-494
4,4-Dimethyl-2-pentyne ≥98%
4,4-Dimethyl-2-pentyne
Formula:
C₇H₁₂ MW: 96.17 g/mol Boiling Pt: 83 °C (1013 hPa) Melting Pt: -82.4 °C Density: 0.7176 g/cm³ (20 °C) Flash Pt: 14 °F |
MDL Number:
MFCD00041613 CAS Number: 999-78-0 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...