Boron ≥99.99%, crystalline powder

Supplier: STREM CHEMICALS INC MS
Warning

05-0065-250MG 05-0065-1G
100204-768EA 141.54 USD
100204-768 100202-592
Boron ≥99.99%, crystalline powder
Boron
Formula: B
MW: 10.81 g/mol
Boiling Pt: 2550 °C
Melting Pt: 2300 °C
Density: 2.34…2.37
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1 III
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR