Tetrabutylammonium fluoride 1M in THF
Supplier: STREM CHEMICALS INC MS
|
Danger
|
02-5012-250MMOL
02-5012-50MMOL
100200-818EA
336.86
USD
100200-818
100207-510
Tetrabutylammonium fluoride 1M in THF
Tetrabutylammonium fluoride
Formula:
C₁₆H₃₆FN MW: 261.47 g/mol Flash Pt: 1 °F |
MDL Number:
MFCD00011747 CAS Number: 429-41-4 EINECS: 207-057-2 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...