Ammonium iodide ≥99% ACS
Supplier: Spectrum Chemical Mfg. Corp.
|
Warning
|
Ammonium Iodide, Reagent, ACS is used in photographic chemicals and some medications.
As a Reagent ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
NH₄I Melting Pt: 551 °C Density: 2.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011424 CAS Number: 12027-06-4 EINECS: 234-717-7 Merck Index: 13,00531 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...