Aluminium metal, powder 200 Mesh

Supplier: Spectrum Chemical Mfg. Corp.
Danger

A1050-25GM A1050-12KG A1050-2.5KG A1050-500GM
700013-298EA 130.2 USD
700013-298 700013-294 700013-296 700013-300
Aluminium metal, powder 200 Mesh
Aluminium

Aluminum Metal, 200 Mesh, Powder is a soft ductile metal and considered the most common metal in the Earth’s crust. Bauxite is the chief ore of aluminum. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: Al
Boiling Pt: 2057 °C (1013 hPa)
Melting Pt: 659.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3,II

Order Now

Specification Test Results

Particle size 200 Mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR