Copper (I) sulfide ≥99.5% (Cu basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Dicopper sulphide, Chacolcite, Copper(i)sulphide

93-2960-250G 93-2960-50G
100202-058EA 413.29 USD
100202-058 100202-060
Copper (I) sulfide ≥99.5% (Cu basis)
Copper (I) sulphide
Formula: Cu₂S
MW: 159.16 g/mol
Melting Pt: 1100 °C
Density: 5.783 g/cm³ (25 °C)
MDL Number: MFCD00016062
CAS Number: 22205-45-4
EINECS: 244-842-9
Merck Index: 13,02697

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR