Copper(II) sulfate, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cupric sulphate

93-2957-250G 93-2957-50G
100204-700EA 117.48 USD
100204-700 100197-806
Copper(II) sulfate, anhydrous ≥98%
Copper(II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200 °C
Density: 3.603 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,02722

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR