Copper(II) perchlorate hexahydrate 98
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cupric perchlorate hexahydrate
93-2947-25G
93-2947-100G
100197-804EA
36.8
USD
100197-804
100202-054
Copper(II) perchlorate hexahydrate 98
Copper(II) perchlorate hexahydrate
Formula:
Cl₂CuO₈·6H₂O MW: 370.54 Boiling Pt: 120 °C (dec.) Melting Pt: 82 Density: 2.225 |
MDL Number:
MFCD00149666 CAS Number: 10294-46-9 EINECS: 237-391-4 UN: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...