Copper(I) oxide ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cuprous oxide, Dicopper oxide
93-2944-250G
93-2944-1000G
100205-494EA
42.46
USD
100205-494
100204-292
Copper(I) oxide ≥95%
Copper(I) oxide
Formula:
Cu₂O MW: 143.09 g/mol Boiling Pt: 1800 °C (1013 hPa) Melting Pt: 1235 °C Density: 5.7 to 6.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010974 CAS Number: 1317-39-1 EINECS: 215-270-7 UN: 3077 ADR: 9,III Merck Index: 13,02694 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...