Copper(II) nitrate trihydrate ≥99.999% (Cu basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate

93-2940-100G 93-2940-5G 93-2940-25G
100197-216EA 331.2 USD
100197-216 100210-340 100207-018
Copper(II) nitrate trihydrate ≥99.999% (Cu basis), PURATREM
Copper(II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Melting Pt: 114.5 °C
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 10031-43-3
EINECS: 221-838-5
UN: 1477
ADR: 5.1,III
Merck Index: 13,02671

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR