Copper(II) nitrate trihydrate ≥99.5%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate

93-2939-250G 93-2939-1000G
100199-064EA 48.12 USD
100199-064 100208-148
Copper(II) nitrate trihydrate ≥99.5%
Copper(II) nitrate trihydrate
Formula: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Melting Pt: 114.5 °C
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 10031-43-3
EINECS: 221-838-5
UN: 1477
ADR: 5.1,III
Merck Index: 13,02671

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR