Copper(I) iodide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cuprous iodide

93-2936-100G 93-2936-500G
100196-598EA 58.03 USD
100196-598 100207-830
Copper(I) iodide ≥98%
Copper(I) iodide
Formula: CuI
MW: 190.45 g/mol
Boiling Pt: 1290 °C
Melting Pt: 605 °C
Density: 5.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR