Copper(II) fluoride, anhydrous ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cupric fluoride
93-2923-5G
93-2923-25G
100204-290EA
48.12
USD
100204-290
100202-052
Copper(II) fluoride, anhydrous ≥99%
Copper(II) fluoride
Formula:
CuF₂ MW: 101.54 Boiling Pt: 950 (dec.) Melting Pt: 855 Density: 4.23 |
MDL Number:
MFCD00010977 CAS Number: 7789-19-7 EINECS: 232-147-3 UN: 1759 ADR: 8 III Merck Index: 12,02704 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...