Copper(II) chloride dihydrate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

93-2913-1000G 93-2913-250G
100210-866EA 461.42 USD
100210-866 100209-430
Copper(II) chloride dihydrate ≥99% ACS
Copper(II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR