Copper(II) chloride, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Copper dichloride, Cupric chloride

93-2912-100G 93-2912-500G
100207-010EA 42.46 USD
100207-010 100196-594
Copper(II) chloride, anhydrous ≥98%
Copper(II) chloride
Formula: Cl₂Cu
MW: 134.45 g/mol
Boiling Pt: 1365.85
Melting Pt: 628
Density: 3.386
Storage Temperature: Ambient
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 13,02660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR