Nickel (II) oxide, black ≥99.9%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

93-2861-500G 93-2861-100G
100197-214EA 155.69 USD
100197-214 100207-826
Nickel (II) oxide, black ≥99.9%
Nickel (II) oxide
Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1955 °C
Density: 6.72 g/cm³ (20 °C)
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7
Merck Index: 13,06540

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR