Nickel(II) sulphate hexahydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickelous sulphate, Hexahydrate, Nickel sulphate hexahydrate

93-2839-2000G 93-2839-500G
100203-434EA 467.08 USD
100203-434 100204-284
Nickel(II) sulphate hexahydrate ≥98% ACS
Nickel(II) sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840
Density: 2.07
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1 III
Merck Index: 13,06541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR