Nickel (II) oxide ≥99.99% (Ni basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

93-2833-25G 93-2833-100G
100199-540EA 171.26 USD
100199-540 100207-824
Nickel (II) oxide ≥99.99% (Ni basis), PURATREM
Nickel (II) oxide
Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1990 °C
Density: 4.6 to 6.67 g/cm³ (20 °C)
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR