Nickel (II) oxide, green ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

93-2832-50G 93-2832-250G
100209-068EA 86.34 USD
100209-068 100206-022
Nickel (II) oxide, green ≥99%
Nickel (II) oxide
Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1990 °C
Density: 6.67
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7
Merck Index: 13,06536

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR