Nickel diiodide, anhydrous ≥99.5% (Ni basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel (II) iodide, Nickel iodide (NiI2)

93-2827-50G 93-2827-10G
100199-048EA 570.4 USD
100199-048 100204-282
Nickel diiodide, anhydrous ≥99.5% (Ni basis)
Nickel diiodide
Formula: I₂Ni
Melting Pt: 797 °C
Density: 5.83 g/cm³ (20 °C)
MDL Number: MFCD00016264
CAS Number: 13462-90-3
EINECS: 236-666-6
UN: 3260
ADR: 8,II
Merck Index: 12,06594

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR