Nickel(II) fluoride tetrahydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel difluoride tetrahydrate

93-2819-25G 93-2819-100G
100205-478EA 42.46 USD
100205-478 100208-144
Nickel(II) fluoride tetrahydrate ≥98%
Nickel(II) fluoride tetrahydrate
Formula: F₂Ni·4H₂O
MW: 168.75 g/mol
MDL Number: MFCD00016263
CAS Number: 13940-83-5
EINECS: 233-071-3
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR