Nickel(II) fluoride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-2818-10G 93-2818-50G
100197-792EA 99.08 USD
100197-792 100201-272
Nickel(II) fluoride, anhydrous ≥99%
Nickel(II) fluoride
Formula: F₂Ni
Melting Pt: 1360…1380 °C
Density: 4.63
MDL Number: MFCD00016263
CAS Number: 10028-18-9
EINECS: 233-071-3
UN: 3288
ADR: 6.1,III
Merck Index: 12,06591

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR