Nickel(II) chloride, anhydrous ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Nickel dichloride
93-2809-50G
93-2809-250G
100202-036EA
92
USD
100202-036
100210-686
Nickel(II) chloride, anhydrous ≥98%
Nickel(II) chloride
Formula:
Cl₂Ni MW: 129.6 g/mol Melting Pt: 1001 °C Density: 3.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011142 CAS Number: 7718-54-9 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 12,06588 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...