Cobalt ≥99.8%, Powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2761-500G
93-2761-100G
93-2761-25G
100200-102EA
607.2
USD
100200-102
100203-432
100204-276
Cobalt ≥99.8%, Powder
Cobalt
Formula:
Co Boiling Pt: 2870 °C Melting Pt: 1495 °C Density: 8,9 |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...