Cobalt ≥99.8%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-2760-50G 93-2760-250G
100205-476EA 77.85 USD
100205-476 100203-430
Cobalt ≥99.8%, powder
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 II
Merck Index: 12,02488

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR