Cobalt(II) thiocyanate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cobalt dirhodanide, Cobaltous rhodanide, Cobalt(II) thiocyanate, Cobalt dithiocyanate, Cobaltous thiocyanate
93-2753-10G
93-2753-50G
100197-788EA
63.69
USD
100197-788
100209-064
Cobalt(II) thiocyanate ≥95%
Cobalt (II) thiocyanate
Formula:
C₂CoN₂S₂ MW: 175.1 g/mol Density: 1.955 g/cm³ (25 °C) |
MDL Number:
MFCD00010937 CAS Number: 3017-60-5 EINECS: 221-156-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...