Cobalt sulfide ≥99.5% (Co basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2751-25G
93-2751-5G
100210-320EA
179.75
USD
100210-320
100205-474
Cobalt sulfide ≥99.5% (Co basis)
Cobalt sulphide
Formula:
CoS MW: 91 g/mol Melting Pt: 1195 °C Density: 5.45 g/cm³ (20 °C) |
MDL Number:
MFCD00016032 CAS Number: 1317-42-6 EINECS: 215-273-3 Merck Index: 12,02511 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...