Cobalt(II) sulfate heptahydrate ≥99.999% (Co basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cobalt sulphate heptahydrate, Cobaltous sulphate heptahydrate

93-2750-50G 93-2750-10G
100206-020EA 585.97 USD
100206-020 100205-472
Cobalt(II) sulfate heptahydrate ≥99.999% (Co basis), PURATREM
Cobalt(II) sulphate heptahydrate
Formula: CoSO₄·7H₂O
MW: 281.1 g/mol
Melting Pt: 96.8 °C
Density: 2.03 g/cm³ (20 °C)
MDL Number: MFCD00149658
CAS Number: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck Index: 12,02510

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR