Cobalt(II) perchlorate hexahydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cobalt diperchlorate hexahydrate
93-2747-250G
93-2747-50G
100209-062EA
525.11
USD
100209-062
100197-786
Cobalt(II) perchlorate hexahydrate ≥98%
Cobalt(II) perchlorate hexahydrate
Formula:
Cl₂CoO₈·6H₂O MW: 365.93 g/mol |
MDL Number:
MFCD00150207 CAS Number: 13478-33-6 EINECS: 236-653-5 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...