Cobalt(II) nitrate hexahydrate ≥99.999% (Co basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

93-2746-25G 93-2746-5G
100199-276EA 298.65 USD
100199-276 100200-546
Cobalt(II) nitrate hexahydrate ≥99.999% (Co basis), PURATREM
Cobalt(II) nitrate hexahydrate
Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 55…56 °C
Density: 1.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 13,02469

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR