Cobalt(II) iodide, anhydrous ≥95% (99.5%-Co)
Supplier: STREM CHEMICALS INC MS
93-2743-10G
93-2743-50G
100200-100EA
174.09
USD
100200-100
100205-468
Cobalt(II) iodide, anhydrous ≥95% (99.5%-Co)
Cobalt(II) iodide
Formula:
CoI₂ MW: 312.74 g/mol Boiling Pt: 570 °C (1013 hPa) Melting Pt: 515 °C Density: 5.68 g/cm³ (20 °C) |
MDL Number:
MFCD00016030 CAS Number: 15238-00-3 EINECS: 239-283-2 Merck Index: 12,02504 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...