Cobalt(III) fluoride ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cobaltic fluoride, Cobalt trifluoride

93-2740-10G 93-2740-50G
100202-028EA 100.49 USD
100202-028 100202-446
Cobalt(III) fluoride ≥99%
Cobalt(III) fluoride
Formula: CoF₃
MW: 115.93 g/mol
Melting Pt: 927 °C
Density: 3.88 g/cm³ (20 °C)
MDL Number: MFCD00010942
CAS Number: 10026-18-3
EINECS: 233-062-4
UN: 1759
ADR: 8,II
Merck Index: 12,02491

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR