Cobalt(II) fluoride, anhydrous ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cobalt difluoride, Cobaltous fluoride
93-2738-50G
93-2738-10G
100210-318EA
237.78
USD
100210-318
100200-098
Cobalt(II) fluoride, anhydrous ≥99%
Cobalt(II) fluoride
Formula:
CoF₂ MW: 96.93 Boiling Pt: 1400 Melting Pt: 1200 Density: 4.46 |
MDL Number:
MFCD00010941 CAS Number: 10026-17-2 EINECS: 233-061-9 UN: 2923 ADR: 8 II Merck Index: 13,02465 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...