Cobalt(II) chloride hexahydrate ≥99.999% (Co basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

93-2735-25G 93-2735-5G
100206-988EA 331.2 USD
100206-988 100199-038
Cobalt(II) chloride hexahydrate ≥99.999% (Co basis), PURATREM
Cobalt(II) chloride hexahydrate
Formula: CoCl₂·6H₂O
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.924
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR