Cobalt(II) carbonate hydrate ≥99% (Co basis) (Co min. 45.5%)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2733-1000G
93-2733-50G
93-2733-250G
100207-284EA
366.58
USD
100207-284
100205-466
100210-316
Cobalt(II) carbonate hydrate ≥99% (Co basis) (Co min. 45.5%)
Cobalt(II) carbonate hydrate
Formula:
CCoO₃·H₂O MW: 136.96 g/mol Density: 4,13 |
MDL Number:
MFCD00149654 CAS Number: 57454-67-8 EINECS: 208-169-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...