Cobalt(II) chloride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-2721-25G 93-2721-100G
100204-270EA 63.69 USD
100204-270 100205-464
Cobalt(II) chloride, anhydrous ≥99%
Cobalt(II) chloride
Formula: Cl₂Co
Boiling Pt: 1049 °C
Melting Pt: 724 °C
Density: 3.356
Storage Temperature: Refrigerator
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,02498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR