Hexaamminecobalt trichloride ≥99.999% (Co basis), PURATREM
Supplier: STREM CHEMICALS INC MS
93-2708-10G
93-2708-50G
100199-036EA
94.83
USD
100199-036
100207-282
Hexaamminecobalt trichloride ≥99.999% (Co basis), PURATREM
Hexaamminecobalt trichloride
Formula:
H₁₈CoCl₃N₆ Melting Pt: 217 °C Density: 1.710 |
MDL Number:
MFCD00036304 CAS Number: 10534-89-1 EINECS: 234-103-9 Merck Index: 12,04711 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...