Cobalt (III) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Cobalt (III) tris(2,4-pentanedionato-O,O'), Cobalt (III) 2,4-pentanedionate, Cobalt tris(acetylacetonate), Tris(2,4-pentanedionato-O,O')cobalt (III)

93-2702-10G 93-2702-250G 93-2702-50G
100199-034EA 45.29 USD
100199-034 100196-578 100196-580
Cobalt (III) acetylacetonate ≥98%
Cobalt (III) acetylacetonate
Formula: C₁₅H₂₁CoO₆
MW: 356.26 g/mol
Boiling Pt: 150 °C (1 mmHg) (subl.)
Melting Pt: 216 °C
MDL Number: MFCD00013488
CAS Number: 21679-46-9
EINECS: 244-527-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR