Iron(II) fluoride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferrous fluoride, Iron difluoride

93-2675-25G 93-2675-5G
100204-264EA 206.65 USD
100204-264 100204-266
Iron(II) fluoride, anhydrous ≥99%
Iron(II) fluoride
Formula: F₂Fe
MW: 93.84 g/mol
Boiling Pt: 1100 °C (1013 hPa)
Melting Pt: > 1000 °C
Density: 4.09 g/cm³ (20 °C)
MDL Number: MFCD00016093
CAS Number: 7789-28-8
EINECS: 232-155-7
UN: 3288
ADR: 6.1,III
Merck Index: 12,04093

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR