Iron(III) nitrate nonahydrate ≥99.999% (Fe basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

93-2672-25G 93-2672-5G
100200-092EA 556.25 USD
100200-092 100206-984
Iron(III) nitrate nonahydrate ≥99.999% (Fe basis), PURATREM
Iron(III) nitrate nonahydrate
Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 °C (dec.)
Melting Pt: 47.2 °C
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 13,04057

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR