Stainless steel, foil type 304
Supplier: STREM CHEMICALS INC MS
93-2666-1500MM
93-2666-300MM
100210-304EA
243.45
USD
100210-304
100196-576
Stainless steel, foil type 304
Stainless steel
Formula:
Fe Boiling Pt: 3000 °C Melting Pt: 1535 °C Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00148499 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...