Iron(III) bromide, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ferric bromide, Iron tribromide

93-2665-10G 93-2665-50G
100207-814EA 100.49 USD
100207-814 100207-816
Iron(III) bromide, anhydrous ≥99%
Iron(III) bromide
Formula: Br₃Fe
MW: 295.56 g/mol
Melting Pt: Dec.
MDL Number: MFCD00016082
CAS Number: 10031-26-2
EINECS: 233-089-1
UN: 3260
ADR: 8 III
Merck Index: 12,04060

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR